Skip to Content
Merck
All Photos(1)

Documents

33670

Supelco

BDE No 47 solution

50 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,4,4′-Tetrabromodiphenyl ether solution, 2,2′,4,4′-TetraBDE, PBDE 47

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Br4O
CAS Number:
Molecular Weight:
485.79
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Brc1ccc(Oc2ccc(Br)cc2Br)c(Br)c1

InChI

1S/C12H6Br4O/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6H

InChI key

XYBSIYMGXVUVGY-UHFFFAOYSA-N

Application

BDE No 47 may be used as a reference standard for the identification of BDE No 47 congener in technical flame-retardant, Bromkal 70-5DE using gas chromatography technique(GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinli Xiong et al.
Ecotoxicology and environmental safety, 164, 164-171 (2018-08-15)
Polybrominated diphenyl ethers (PBDEs) are extremely incessant anthropogenic contaminants found in the environment, with dreadful risk to aquatic ecosystems. However, there is a limited amount of data concerning their impacts on freshwater organisms. 2,2',3,3',4,4',5,5',6,6'-decabromodiphenyl ether (BDE-209) and 2,2',4,4'-tetrabromodiphenyl ether (BDE-47)
Yanhua Liu et al.
Environmental pollution (Barking, Essex : 1987), 258, 113720-113720 (2019-12-14)
As a persistent organic pollutant, 2,2',4,4'-tetrabromodiphenyl ether (BDE-47) has been widely detected in aquatic environments. However, studies on the fate and transfer of BDE-47 in the aquatic food chain remain scarce. In this study, we investigated the bioaccumulation and elimination
Yucheng Zhao et al.
Ecotoxicology and environmental safety, 176, 279-287 (2019-04-05)
2,2',4,4'-tetrabrominated diphenyl ether (BDE-47) and 2,2',4,4',5-pentabromodiphenyl ether (BDE-99) are two typical polybrominated diphenyl ethers (PBDEs), and studies have proven that these PBDs can disrupt the behaviors and physical function of aquatic organisms. However, little is known about the compositional impacts
Gas chromatographic identification and quantification of polybrominated diphenyl ethers in a commercial product, Bromkal 70-5DE.
Sjodin.A, et al.
Journal of Chromatography A, 822, 83-89 (1998)
Youssef Oulhote et al.
Environment international, 119, 79-88 (2018-06-26)
Prenatal exposure to polybrominated diphenyl ethers (PBDEs) has been associated with cognitive deficits and behavioral problems in children. To date, no study has examined this exposure in association with neurobehavioral development in infants younger than 12 months assessed with observational tasks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service