Skip to Content
Merck
All Photos(1)

Key Documents

33663

Supelco

BDE No 28 solution

50 μg/mL in isooctane, analytical standard

Synonym(s):

2,4,4′-Tribromodiphenyl ether solution, 2,4,4′-TriBDE, PBDE 28

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$728.00

$728.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$728.00

About This Item

Empirical Formula (Hill Notation):
C12H7Br3O
CAS Number:
Molecular Weight:
406.90
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$728.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Brc1ccc(Oc2ccc(Br)cc2Br)cc1

InChI

1S/C12H7Br3O/c13-8-1-4-10(5-2-8)16-12-6-3-9(14)7-11(12)15/h1-7H

InChI key

UPNBETHEXPIWQX-UHFFFAOYSA-N

Related Categories

General description

BDE No 28 is a ubiquitous environmental contaminant with low acute toxicity, belonging to the class of polybrominated diphenyl ethers.[1] It can find wide applications as a flame-retardant additive in consumer products ranging from textiles to polymers.

Application

BDE No 28 may be used as an analytical reference standard for the determination of the analyte in fish samples using gas chromatography coupled to mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Arioli et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 36(8), 1244-1252 (2019-06-14)
The exposure to several compounds such as polychlorinated biphenyls (PCBs), organochlorine pesticides (OCPs), organophosphorus pesticides (OPs), polycyclic aromatic hydrocarbon (PAHs), perfluoroalkyl substances (PFASs) and polybrominated diphenyl ethers (PBDEs) is a public health issue. The European Union (EU) recommended that its
GC / MS analysis of polybrominated diphenyl ethers in fish collected from the Inland Sea of ??Seto, Japan.
Akutsu K, et al.
Chemosphere, 44(6), 1325-1333 (2001)
Anders Larsson et al.
Journal of medicinal chemistry, 49(25), 7366-7372 (2006-12-08)
To investigate androgen receptor (AR) activation by exogenous compounds, we used a combination of experimental analysis and theoretical modeling to compare a set of brominated flame retardants (BFRs) to dihydrotestosterone (DHT) with regard to ligand docking, AR binding, and AR
Luca Maria Chiesa et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(7), 1340-1355 (2018-03-14)
Demand for honey is increasing, especially if it is organic and if its nutritional properties are linked to untreated environments in order to guarantee quality for health. Sources of contamination of honey can be divided into environmental and apicultural. Therefore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service