Skip to Content
Merck
All Photos(1)

Key Documents

33563

Supelco

4-tert-Octylphenol-monoethoxylate-ring-13C6 solution

10 μg/mL in acetone, analytical standard

Synonym(s):

2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethanol-13C6, 4-tert-OP1EO-13C6

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$564.00

$564.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$564.00

About This Item

Empirical Formula (Hill Notation):
13C6C10H26O2
CAS Number:
Molecular Weight:
256.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$564.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+6

storage temp.

−20°C

SMILES string

CC(C)(C)CC(C)(C)[13c]1[13cH][13cH][13c](OCCO)[13cH][13cH]1

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3/i6+1,7+1,8+1,9+1,13+1,14+1

InChI key

JYCQQPHGFMYQCF-FQPQTBQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

4-tert-Octylphenol-monoethoxylate-ring-13C6 solution is an analytical standard used for the analysis of mono and di-ethoxylates of nonylphenol (NP) and octylphenol (OP) in environmental samples using solid-phase micro extraction followed by gas chromatography with mass spectrometry (SPME-GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service