Recommended Products
grade
technical grade
Quality Level
Assay
~97% (HPLC)
form
powder
mp
>300 °C (lit.)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[O-][N+](=O)c1ccc(cc1)N(c2ccc(cc2)[N+]([O-])=O)c3ccc(cc3)[N+]([O-])=O
InChI
1S/C18H12N4O6/c23-20(24)16-7-1-13(2-8-16)19(14-3-9-17(10-4-14)21(25)26)15-5-11-18(12-6-15)22(27)28/h1-12H
InChI key
LSNJBIDKQIRWRQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service