Skip to Content
Merck
All Photos(1)

Key Documents

33483

Sigma-Aldrich

4,4′-Diazido-2,2′-stilbenedisulfonic acid disodium salt tetrahydrate

≥99.0%

Synonym(s):

Disodium 4,4′-diazidostilbene-2,2′-disulfonate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N6Na2O6S2 · 4H2O
CAS Number:
Molecular Weight:
538.42
EC Number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

SMILES string

O.O.O.O.[Na+].[Na+].[O-]S(=O)(=O)c1cc(ccc1\C=C\c2ccc(cc2S([O-])(=O)=O)N=[N+]=[N-])N=[N+]=[N-]

InChI

1S/C14H10N6O6S2.2Na/c15-19-17-11-5-3-9(13(7-11)27(21,22)23)1-2-10-4-6-12(18-20-16)8-14(10)28(24,25)26;;/h1-8H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b2-1+;;

InChI key

HYYYTIDWAIQGHI-SEPHDYHBSA-L

Other Notes

A reagent for enzyme immobilisation on fibroin by photo-crosslinking. The reagent is water-soluble over a wide pH range[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Speth et al.
Archives of biochemistry and biophysics, 275(1), 202-214 (1989-11-15)
The effect of the photoactivated reagent 4,4'-diazidostilbene 2,2'-disulfonic acid (DASS) on rat liver microsomal glucose-6-phosphatase has been investigated in order to analyze the accessibility and the chemical nature of functional sites of the integral enzyme protein. The following results were
Lizhang Wang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 62(11), 2669-2677 (2010-11-26)
Experiments were performed to reduce chemical oxygen demand (COD) from 4,4'-diaminostilbene-2,2'-disulfonic (DSD) acid manufacturing wastewater using electrochemical oxidation coupled with adsorption by granular activated carbon. The COD removal is affected by the residence time and applied voltage. When the residence
Application of photochemical reactions of bis-azido compounds to preparation of an enzyme-polymer film.
S Miyairi et al.
Analytical biochemistry, 97(2), 320-322 (1979-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service