Skip to Content
Merck
All Photos(1)

Key Documents

33180

Sigma-Aldrich

1,8-Diaminonaphthalene

technical, ≥95.0% (HPLC)

Synonym(s):

1,8-Naphthalenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(NH2)2
CAS Number:
Molecular Weight:
158.20
Beilstein:
742841
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥95.0% (HPLC)

form

solid

bp

205 °C/12 mmHg (lit.)

mp

60-65 °C (lit.)
60-65 °C

SMILES string

Nc1cccc2cccc(N)c12

InChI

1S/C10H10N2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H,11-12H2

InChI key

YFOOEYJGMMJJLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>392.0 °F - closed cup

Flash Point(C)

> 200 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohan N Patel et al.
Bioorganic & medicinal chemistry, 17(15), 5648-5655 (2009-07-08)
Here in we tried to increase an antibacterial activity of ciprofloxacin drug due to formation of mixed-ligand complexes. Synthesized compounds were found to be more potent compare to drugs, ligands and metal salt against selective gram((+ve)) and gram((-ve)) organisms. Interaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service