Skip to Content
Merck
All Photos(1)

Key Documents

32924

Supelco

Oxibendazole

VETRANAL®, analytical standard

Synonym(s):

Methyl (5-propoxy-1H-benzimidazol-2-yl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$372.00

$372.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$372.00

About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$372.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CCCOc1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O3/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

RAOCRURYZCVHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxibendazole is an analog of albendazole, that shows activity against Necator, a group of hookworms.[1]
Sigma-Aldrich offers oxibendazole for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyses.

Application

Oxibendazole may be used as an analytical reference standard for the determination of the analyte in dairy samples,[2][3] animal muscle[4] and water samples[5] by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion-pair isolation and liquid chromatographic determination of albendazole, oxfendazole, oxibendazole, and thiabendazole residues in milk
Nakos DS, et al.
Journal of Liquid Chromatography and Related Technologies, 17(19), 4145-4155 (1994)
Human gastrointestinal helminth infections: are they now neglected diseases?
Horton J.
Trends in Parasitology, 19(11), 527-531 (2003)
Determination of benzimidazole residues using liquid chromatography and tandem mass spectrometry
Balizs G
Journal of Chromatography. B, Biomedical Sciences and Applications, 727(1-2), 167-177 (1999)
Development and validation of a liquid chromatographic-electrospray tandem mass spectrometric multiresidue method for anthelmintics in milk
De Ruyck H, et al.
Journal of Chromatography A, 976(1-2), 181-194 (2002)
Analysis of anthelmintics in surface water by ultra high performance liquid chromatography coupled to quadrupole linear ion trap tandem mass spectrometry
Zrncic M, et al.
Chemosphere, 99(1-2), 224-232 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service