Skip to Content
Merck
All Photos(1)

Key Documents

32898

Supelco

Nonylphenol diethoxylate solution

50 μg/mL in acetone, technical, analytical standard

Synonym(s):

NP-2EO technical

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$310.00

$310.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$310.00

About This Item

Empirical Formula (Hill Notation):
C19H32O3
Molecular Weight:
308.46
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$310.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

analytical standard
technical

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nonylphenol diethoxylate (50 μg/mL in acetone, technical) is classified under the family of alkylphenol ethoxylate surfactants majorly used for households, industry and agriculture.[1]

Application

Nonylphenol diethoxylate (50 μg/mL in acetone, technical) may be utilized as an analytical reference standard for the quantification of the analyte in:
  • Fish and shellfish using high performance liquid chromatography-fluorescence detection (HPLC-FLD).[2]
  • Wastewater and sewage sludge using gas chromatography-mass spectrometry (GC-MS).[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of nonylphenol ethoxylates in the aquatic environment by normal phase liquid chromatography-electrospray mass spectrometry.
Shao B, et al.
Journal of Chromatography A, 950(1-2), 167-174 (2002)
Determination of 4-nonylphenol, nonylphenol monoethoxylate, nonylphenol diethoxylate and other alkylphenols in fish and shellfish by high-performance liquid chromatography with fluorescence detection.
Tsuda T, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 746(2), 305-309 (2000)
Simultaneous determination of the endocrine disrupting compounds nonylphenol, nonylphenol ethoxylates, triclosan and bisphenol A in wastewater and sewage sludge by gas chromatography-mass spectrometry.
Gatidou G, et al.
Journal of Chromatography A, 1138(1-2), 32-41 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service