Skip to Content
Merck
All Photos(2)

Key Documents

32853

Supelco

Crystal Violet-d6 trihydrate

analytical standard

Synonym(s):

Basic Violet 3-d6, Methyl Violet 10B-d6

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$586.00

$586.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$586.00

About This Item

Empirical Formula (Hill Notation):
C25D6H24ClN3 · 3H2O
Molecular Weight:
468.06
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$586.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+6

SMILES string

O.O.O.[Cl-].[2H]C([2H])([2H])N(c1ccc(cc1)C(\c2ccc(cc2)N(C)C)=C3/C=CC(\C=C3)=[N+](\C)C)C([2H])([2H])[2H]

InChI

1S/C25H30N3.ClH.3H2O/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;;;;/h7-18H,1-6H3;1H;3*1H2/q+1;;;;/p-1/i1D3,2D3;;;;

InChI key

KFXQTVCGYUQQCQ-WMCWWHCESA-M

Application

Crystal Violet-d6 trihydrate may be used as a precursor for the preparation of crystal violet-d6 which may be used as an internal standard for the determination of crystal violet in aquaculture products by liquid chromatography-tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Hurtaud-Pessel et al.
Journal of chromatography. A, 1218(12), 1632-1645 (2011-02-12)
A method is described for the identification and the quantitative determination of the triphenylmethane dyes, malachite green (MG), crystal violet (CV), brilliant green (BG) and leuco malachite green (LMG) and leuco crystal violet (LCV). The analytes were isolated from the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service