Skip to Content
Merck
All Photos(1)

Key Documents

32741

Sigma-Aldrich

3,3′-Diaminobenzidine

purum, ≥98.0% (CHN)

Synonym(s):

3,3′,4,4′-Biphenyltetramine, 3,3′,4,4′-Tetraaminobiphenyl, DAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2)2C6H3C6H3(NH2)2
CAS Number:
Molecular Weight:
214.27
Beilstein:
1212988
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (CHN)

form

solid

mp

173-176 °C
175-177 °C (lit.)

solubility

dioxane: soluble 0.1 g/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

NC1=C(N)C=CC(C2=CC(N)=C(N)C=C2)=C1

InChI

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

InChI key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photooxidation of 3,3′-diaminobenzidine by Chlamydomonas reinhardii and blue-green algae has been investigated.[1] Electropolymerization of 3,3′-diaminobenzidine on solid substrate electrode (Platinum, Gold and Glassy Electrode) affords stable and water insoluble films.[2]

Application

3,3′-Diaminobenzidine, a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue.[3] 3,3′-Diaminobenzidine may be used for the determination of selenium[4] and for the synthesis of a useful, linear, Schiff-base coordination polymer.[5]
Peroxidase substrate; reagent for spectrophotometric determination of selenium.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photooxidation of 3,3'-diaminobenzidine by blue-green algae and Chlamydomonas reinhardii.
N H Chua
Biochimica et biophysica acta, 267(1), 179-189 (1972-04-20)
Jimmy C Azar et al.
Computational and mathematical methods in medicine, 2014, 647273-647273 (2014-07-26)
Comparing staining patterns of paired antibodies designed towards a specific protein but toward different epitopes of the protein provides quality control over the binding and the antibodies' ability to identify the target protein correctly and exclusively. We present a method
Poly (3, 3'-diaminobenzidine) film on a gold electrode for selective preconcentration and stripping analysis of selenium (IV).
Cai Q and Khoo SB.
Analytical Chemistry, 66(24), 4543-4550 (1994)
Synthesis and characterization of the thermally stable copolymer of tetrakis (salicylaldehydato-O, O') zirconium (IV) and 3, 3'-diaminobenzidine.
Archer RD and Wang B.
Inorganic Chemistry, 29(1), 39-39 (1990)
Determination of traces of selenium 3, 3-diaminobenzidine as selenium (lV) organic reagent.
Cheng K L.
Analytical Chemistry, 28(11), 1738-1742 (1956)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service