Skip to Content
Merck
All Photos(2)

Key Documents

32671

Supelco

Carnidazole

VETRANAL®, analytical standard

Synonym(s):

[2-(2-Methyl-5-nitro-imidazol-1-yl)ethyl]thiocarbamic acid O-methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$572.00

$572.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$572.00

About This Item

Empirical Formula (Hill Notation):
C8H12N4O3S
CAS Number:
Molecular Weight:
244.27
Beilstein:
620260
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$572.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

COC(=S)NCCn1c(C)ncc1[N+]([O-])=O

InChI

1S/C8H12N4O3S/c1-6-10-5-7(12(13)14)11(6)4-3-9-8(16)15-2/h5H,3-4H2,1-2H3,(H,9,16)

InChI key

OVEVHVURWWTPFC-UHFFFAOYSA-N

General description

Carnidazoles belong to the class of nitroimidazole compounds that are generally utilized for the treatment of various diseases in poultry, cattle and pigs.[1]

Application

Carnidazole may be used as an analytical reference standard for the determination of the analyte in:
  • Eggs by liquid chromatography-tandem mass spectrometry (LC-MS/MS).[2]
  • Environmental water samples by dispersive liquid-liquid microextraction (DLLME), followed by micellar electrokinetic chromatography (MEKC).[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Study of carnidazole (R 25831). A new therapeutic agent for vaginal trichomoniasis].
V Fonzé et al.
Revue medicale de Liege, 34(19), 811-815 (1979-10-01)
P Chaudhuri et al.
European journal of obstetrics, gynecology, and reproductive biology, 10(5), 325-328 (1980-06-01)
A double-blind controlled clinical trial of the new, not yet commercially available drug, carnidazole and tinidazole, was carried out in 77 women with a proven vaginal trichomoniasis. 39 patients and their sexual partners received a single dose of 2 g
Novel cation selective exhaustive injection-sweeping procedure for 5-nitroimidazole determination in waters by micellar electrokinetic chromatography using dispersive liquid-liquid microextraction.
Hernandez-Mesa, et al.
Journal of Chromatography A, 1341(46), 65-72 (2014)
L Van Vaeck et al.
Biomedical & environmental mass spectrometry, 16(1-12), 121-129 (1988-10-01)
Laser microprobe mass analysis (LAMMA) was used for the structural characterization of polyfunctional drugs and their oxygenated metabolites, in particular the N-oxides. The spectra usually yield the molecular weight as well as intense fragments. The structural information is characteristically distributed
Rapid confirmatory method for the determination of 11 nitroimidazoles in egg using liquid chromatography tandem mass spectrometry.
Cronly M, et al.
Journal of Chromatography A, 1216(46), 8101-8109 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service