Skip to Content
Merck
All Photos(1)

Key Documents

32635

Supelco

Dimethachlor Metabolite SYN 530561

PESTANAL®, analytical standard

Synonym(s):

2-[(2-Hydroxyacetyl)(2-methoxyethyl)amino]-3-methylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO5
CAS Number:
Molecular Weight:
267.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COCCN(C(=O)CO)c1c(C)cccc1C(O)=O

InChI

1S/C13H17NO5/c1-9-4-3-5-10(13(17)18)12(9)14(6-7-19-2)11(16)8-15/h3-5,15H,6-8H2,1-2H3,(H,17,18)

InChI key

UJKHCLQXFACPDB-UHFFFAOYSA-N

General description

Dimethachlor Metabolite SYN 530561 is one of the metabolites of dimethachlor which is known as a potential pollutant in groundwater.

Application

Dimethachlor Metabolite SYN 530561 may be used as an analytical reference standard for the determination of the analyte in surface water and groundwater samples by direct injection liquid chromatography-electrospray ionization-mass spectrometry (LC-ESI-MS) with multiple reaction monitoring (MRM).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conclusion on the peer review of the pesticide risk assessment of the active substance terbuthylazine.
EFSA Journal, 9(1), 1969-1969 (2011)
A multimethod for the determination of 150 pesticide metabolites in surface water and groundwater using direct injection liquid chromatography-mass spectrometry.
Reemtsma, et al.
Journal of Chromatography A, 1271(1), 95-104 (2013)
Junbing Wu et al.
Cell reports, 28(5), 1167-1181 (2019-08-01)
Parkinson's disease causes the most profound loss of the aldehyde dehydrogenase 1A1-positive (ALDH1A1+) nigrostriatal dopaminergic neuron (nDAN) subpopulation. The connectivity and functionality of ALDH1A1+ nDANs, however, remain poorly understood. Here, we show in rodent brains that ALDH1A1+ nDANs project predominantly to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service