Skip to Content
Merck
All Photos(2)

Key Documents

32567

Supelco

Fenbendazole-d3

VETRANAL®, analytical standard

Synonym(s):

Methyl-d3 5-(phenylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$384.00

$384.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$384.00

About This Item

Empirical Formula (Hill Notation):
C15D3H10N3O2S
CAS Number:
Molecular Weight:
302.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$384.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(=O)Nc1nc2cc(Sc3ccccc3)ccc2[nH]1

InChI

1S/C15H13N3O2S/c1-20-15(19)18-14-16-12-8-7-11(9-13(12)17-14)21-10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)/i1D3

InChI key

HDDSHPAODJUKPD-FIBGUPNXSA-N

General description

Chemical structure: benzimidazole

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2

Target Organs

Liver,lymph node,Stomach,Nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-Young Kim et al.
The Science of the total environment, 579, 940-949 (2016-11-30)
The occurrence trends and effects of 30 human and veterinary pharmaceuticals, including antibiotics, anthelmintics, anti-inflammatory drugs, and β-blockers, in the marine environment, with a focus on seawater, sediment, cultured fish, and their feed collected from coastal and fish farm areas
Ki Yong Kim et al.
Journal of hazardous materials, 400, 123235-123235 (2020-09-20)
The concentrations of 14 perfluoroalkyl substances (PFASs) and 46 pharmaceuticals in raw water and drinking water from five drinking water treatment plants were determined to assess removal of the chemicals during treatment. 10 out of 14 PFASs were detected in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service