Skip to Content
Merck
All Photos(2)

Key Documents

32460

Supelco

Meropenem trihydrate

VETRANAL®, analytical standard

Synonym(s):

(1R,5S,6S)-2-[(3S,5S)-5-(dimethylaminocarbonyl)pyrrolidin-3-ylthio]-6-[(R)-1-hydroxyethyl]-1-methylcarbapen-2-em-3-carboxylic acid trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$184.00

$184.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$184.00

About This Item

Empirical Formula (Hill Notation):
C17H25N3O5S · 3H2O
CAS Number:
Molecular Weight:
437.51
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$184.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

O.O.O.C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(S[C@@H]3CN[C@@H](C3)C(=O)N(C)C)=C(N2C1=O)C(O)=O

InChI

1S/C17H25N3O5S.3H2O/c1-7-12-11(8(2)21)16(23)20(12)13(17(24)25)14(7)26-9-5-10(18-6-9)15(22)19(3)4;;;/h7-12,18,21H,5-6H2,1-4H3,(H,24,25);3*1H2/t7-,8-,9+,10+,11-,12-;;;/m1.../s1

InChI key

CTUAQTBUVLKNDJ-OBZXMJSBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Meropenem trihydrate (Meropenem) is a broad-spectrum carbapenem with antibacterial properties. It inhibits cell wall synthesis in both gram-negative and gram-positive bacteria.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Meropenem has been used as a reference standard in an experiment conducted to investigate the effects on in vivo efficacy caused due to chemical differences in three generic products of meropenem.[1]
Meropenem penetrates cell walls and binds to penicillin-binding protein targets. It acts against aerobes and anaerobes including E. coli, Klebsiella, Clostridium, Enterococcus sp. It is a common cause of mild transient aminotransferase elevations and can rarely result in clinically apparent, cholestatic liver injury.
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Even apparently insignificant chemical deviations among bioequivalent generic antibiotics can lead to therapeutic nonequivalence: the case of meropenem.
Agudelo, M., et al.
Antimicrobial Agents and Chemotherapy, 58 (2), 1005-1018 (2014)
Laurens Manning et al.
PloS one, 9(7), e102023-e102023 (2014-07-16)
Concerns regarding the clinical impact of meropenem instability in continuous infusion (CI) devices may contribute to inconsistent uptake of this method of administration across outpatient parenteral antimicrobial therapy (OPAT) services. We retrospectively reviewed the clinical efficacy and safety of CIs
Ryuichi Fukuda et al.
Developmental cell, 51(1), 62-77 (2019-09-10)
Mechanical forces regulate cell behavior and tissue morphogenesis. During cardiac development, mechanical stimuli from the heartbeat are required for cardiomyocyte maturation, but the underlying molecular mechanisms remain unclear. Here, we first show that the forces of the contracting heart regulate
Johannes Zander et al.
Clinical chemistry and laboratory medicine, 53(5), 781-791 (2014-10-11)
Recent studies have demonstrated highly variable blood concentrations of piperacillin, tazobactam, cefepime, meropenem, ciprofloxacin and linezolid in critically ill patients with a high incidence of sub-therapeutic levels. Consequently, therapeutic drug monitoring (TDM) of these antibiotics has to be considered, requiring

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service