Skip to Content
Merck
All Photos(1)

Documents

32452

Supelco

Dinitolmide

VETRANAL®, analytical standard

Synonym(s):

3,5-Dinitro-o-toluamide, 2-Methyl-3,5-dinitrobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H2(CH3)CONH2
CAS Number:
Molecular Weight:
225.16
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

mp

170-180 °C (lit.)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

Cc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C8H7N3O5/c1-4-6(8(9)12)2-5(10(13)14)3-7(4)11(15)16/h2-3H,1H3,(H2,9,12)

InChI key

ZEFNOZRLAWVAQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dinitolmide is a nitrobenzamide anticoccidial agent widely used in poultry farming because of its low toxicity and stability.

Application

Dinitolmide may be used as an analytical reference standard for the quantification of the analyte in animal tissues poultry, livestock, and aquatic tissues using chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Equilibrium solubility of dinitolmide in several neat solvents and binary aqueous co-solvent mixtures: Experimental determination and thermodynamic analysis
Wu J, et al.
The Journal of Chemical Thermodynamics, 132(3), 373-382 (2019)
Veterinary Pharmacology and Therapeutics (2009)
Multi-residue analysis using liquid chromatography tandem mass spectrometry for detection of 20 coccidiostats in poultry, livestock, and aquatic tissues
Chang HS, et al.
Journal of food and drug analysis, 27(3), 703-716 (2019)
Liqun Wang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(5), 1445-1455 (2016-02-05)
The role that glia play in neurological disease is poorly understood but increasingly acknowledged to be critical in a diverse group of disorders. Here we use a simple genetic model of Alexander disease, a progressive and severe human degenerative nervous
A multi-residue method for 17 anticoccidial drugs and ractopamine in animal tissues by liquid chromatography-tandem mass spectrometry and time-of-flight mass spectrometry.
Matus JL and Boison JO
Drug Testing and Analysis, 8(5-6), 465-476 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service