Skip to Content
Merck
All Photos(2)

Key Documents

32439

Supelco

Nifurpirinol

VETRANAL®, analytical standard

Synonym(s):

Furpirinol, Furpyrinol, {6-[2-(5-Nitro-2-furanyl)vinyl]-2-pyridinyl}methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N2O4
CAS Number:
Molecular Weight:
246.22
Beilstein:
1216943
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

OCc1cccc(C=Cc2ccc(o2)[N+]([O-])=O)n1

InChI

1S/C12H10N2O4/c15-8-10-3-1-2-9(13-10)4-5-11-6-7-12(18-11)14(16)17/h1-7,15H,8H2

InChI key

JQKHJQJVKRFMCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nifurpirinol belongs to the nitrofuran class of compounds.[1]

Application

Nifurpirinol may be used as an analytical reference standard for the determination of the analyte in animal based food products by ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Kimura et al.
National Cancer Institute monograph, 65, 139-154 (1984-05-01)
Chromatophoromas in the croaker nibe, Nibea mitsukurii, are common neoplasms in feral fish which inhabit the shallow water in a unique geographic distribution along the Pacific coast of Japan. We undertook surveys of the epizootiology of tumor-bearing fish at 25
N M Angelini et al.
Revista Argentina de microbiologia, 20(1), 37-48 (1988-01-01)
During intensive culture of South-American catfish, Rhamdia sapo fingerlings an epizootic similar to fin rot was developed. Aeromonas hydrophila and Pseudomonas aeruginosa were isolated and they were treated and controlled with daily short dip, 1 ppm Furanace. The disease was
Acute toxicity of nifurpirinol, a fish chemotherapeutant, to milkfish (Chanos chanos) fingerlings.
C T Tamse et al.
Bulletin of environmental contamination and toxicology, 52(3), 346-350 (1994-03-01)
Y Nakamura et al.
Mutation research, 104(1-3), 61-66 (1982-04-01)
Nifurpirinol, 6-hydroxymethyl-2-[2-(5-nitro-2-furyl)vinyl]pyridine, which has been widely used as an antibacterial drug against diseases of fish, was found to be a potent mutagen. The mutagenic effect was dose-related, and the potencies were calculated to be 1.49 X 10(3), 7.24 X 10(4)
Changes induced in the gills of milkfish (Chanos chanos Forsskål) fingerlings after acute exposure to nifurpirinol (Furanace; P-7138).
C T Tamse et al.
Bulletin of environmental contamination and toxicology, 54(4), 591-596 (1995-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service