Skip to Content
Merck
All Photos(1)

Key Documents

32432

Supelco

Dimefluthrin

PESTANAL®, analytical standard

Synonym(s):

2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl (1RS,3RS;1RS,3SR)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22F4O3
CAS Number:
Molecular Weight:
374.37
Beilstein:
11644422
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COCc1c(F)c(F)c(COC(=O)C2C(\C=C(/C)C)C2(C)C)c(F)c1F

InChI

1S/C19H22F4O3/c1-9(2)6-12-13(19(12,3)4)18(24)26-8-11-16(22)14(20)10(7-25-5)15(21)17(11)23/h6,12-13H,7-8H2,1-5H3

InChI key

OOWCJRMYMAMSOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimefluthrins belong to the pyrethroids class of compounds.[1]

Application

Dimefluthrin may be used as an analytical reference standard for the determination of the analyte in:
  • Indoor air samples by gas chromatography-mass spectrometry (GC/MS) operating in negative chemical ionization (NCI) mode.[1]
  • Agricultural sediment samples by GC/MS.[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrethroids in indoor air during application of various mosquito repellents: Occurrence, dissipation and potential exposure risk.
Li H, et al.
Chemosphere, 144, 2427-2435 (2016)
Huizhen Li et al.
Chemosphere, 144, 2427-2435 (2015-11-30)
Commercial mosquito repellents (MRs) are generally applied as mosquito coils, electric vaporizers (liquid and solid) or aerosol spray, with pyrethroids often being the active ingredients. Four types of MRs were applied individually in a 13-m(2) bedroom to study the occurrence
Legacy and current-use insecticides in agricultural sediments from South China: impact of application pattern on occurrence and risk.
Wei Y, et al.
Journal of Agricultural and Food Chemistry, 65(21), 4247-4254 (2017)
Manas Sarkar et al.
Journal of economic entomology, 111(3), 1376-1381 (2018-04-17)
Intensive exposure to insecticides has resulted in the evolution of insecticide resistance in the mosquitoes. We tested the bio-efficacy of two Culex quinquefasciatus Say (Diptera: Culicidae) laboratory strains differentially bio-responsive to pyrethroids to understand the comparative efficacy of different polyfluorobenzyle

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service