Skip to Content
Merck
All Photos(1)

Key Documents

32428

Supelco

Orysastrobin

PESTANAL®, analytical standard

Synonym(s):

(2E)-2-(Methoxyimino)-2-{2-[(3E,5E,6E)-5-(methoxyimino)-4,6-dimethyl-2,8-dioxa-3,7-diazanona-3,6-dien-1-yl]phenyl}-N-methylacetamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$236.00

$236.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$236.00

About This Item

Empirical Formula (Hill Notation):
C18H25N5O5
CAS Number:
Molecular Weight:
391.42
Beilstein:
11330196
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$236.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)C(=N\OC)\c1ccccc1CO\N=C(C)\C(=N\OC)\C(C)=N\OC

InChI

1S/C18H25N5O5/c1-12(20-25-4)16(22-26-5)13(2)21-28-11-14-9-7-8-10-15(14)17(23-27-6)18(24)19-3/h7-10H,11H2,1-6H3,(H,19,24)/b20-12+,21-13+,22-16+,23-17+

InChI key

JHIPUJPTQJYEQK-ZLHHXESBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Orysastrobin is one of the analogs of strobilurin fungicide.[1][2]

Application

Orysastrobin may be used as an analytical reference standard for the determination of the analyte in:
  • Rice samples by high performance liquid chromatography equipped with ultraviolet (HPLC-UV) detection and LC coupled to tandem mass spectrometry (MS/MS) with electrospray ionization (ESI) and selected reaction monitoring (SRM) detection.[2]
  • Food samples by atmospheric-pressure solids-analysis probe (ASAP)[3] as well as desorption electrospray ionization-mass spectrometry (DESI-MS)[4] techniques.
  • Hot pepper (Capsicum annuum L.) samples by modified QuEChERS (quick, easy, cheap, effective, rugged, and safe) method, followed by analysis using LC-MS/MS with SRM detection.[5]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue method for the determination of 227 pesticides in hot pepper (Capsicum annuum L.) by liquid chromatography with tandem mass spectrometry.
Zhu YZ, et al.
Journal of Separation Science, 37(20), 2947-2954 (2014)
Simultaneous determination of orysastrobin and its isomers in rice using HPLC-UV and LC-MS/MS.
Kwon CH, et al.
Journal of Agricultural and Food Chemistry, 59(20), 10826-10830 (2011)
Quantitative analysis of fungicide azoxystrobin in agricultural samples with rapid, simple and reliable monoclonal immunoassay.
Watanabe E and Miyake S
Food Chemistry, 136(2), 695-702 (2013)
An assessment of atmospheric-pressure solids-analysis probes for the detection of chemicals in food.
Fussell RJ, et al.
TrAC, Trends in Analytical Chemistry, 29(11), 1326-1335 (2010)
Desorption electrospray ionization mass spectrometry in the analysis of chemical food contaminants in food.
Nielen MWF, et al.
TrAC, Trends in Analytical Chemistry, 30(2), 165-180 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service