Skip to Content
Merck
All Photos(1)

Key Documents

32280

Sigma-Aldrich

Diethylene glycol monomethyl ether

purum, ≥99.0% (GC)

Synonym(s):

2-(2-Methoxyethoxy)ethanol, Methyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
120.15
Beilstein:
1697812
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

vapor density

4.14 (vs air)

vapor pressure

0.2 mmHg

grade

purum

Assay

≥99.0% (GC)

expl. lim.

0.34-6.3 % (lit.)

refractive index

n20/D 1.426

bp

190-194 °C (lit.)

mp

-70 °C (lit.)

density

1.020 g/mL at 20 °C (lit.)

SMILES string

COCCOCCO

InChI

1S/C5H12O3/c1-7-4-5-8-3-2-6/h6H,2-5H2,1H3

InChI key

SBASXUCJHJRPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R R Miller et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 5(6 Pt 1), 1174-1179 (1985-12-01)
Male and female Fischer 344 rats were exposed to 0, 30, 100, or 216 ppm diethylene glycol monomethyl ether (DEGME) vapors (0, 0.15, 0.49, or 1.06 mg/liter) 6 hr/day, 5 days/week, for 13 weeks. The 216-ppm exposure level was the
Hao Zhu et al.
Chemical communications (Cambridge, England), 48(96), 11766-11768 (2012-11-02)
Rlyso, a highly selective and sensitive pH sensor, can stain lysosomes with a novel lysosome-locating group, methylcarbitol. Rlyso was successfully used to detect lysosomal pH changes during apoptosis or induced by chloroquine while avoiding the "alkalizing effect" on lysosomes of
B D Hardin et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 6(3), 430-439 (1986-04-01)
Diethylene glycol monomethyl ether (diEGME) was one of 15 glycols tested in CD-1 mice using a short-term in vivo reproductive toxicity assay (Chernoff/Kavlock test). Because results were strongly suggestive of potential reproductive toxicity, a teratology study was conducted in Sprague-Dawley
Induction of r-GTP by ethylene glycol monomethyl ether.
T Kawamoto et al.
Toxicology and industrial health, 7(5-6), 473-478 (1991-09-01)
T Kawamoto et al.
Toxicology, 62(3), 265-274 (1990-06-01)
Glycol ethers have been extensively used in industry over the past 40-50 years. Numerous studies on the toxicity of glycol ethers have been performed, however, the effects of glycol ethers on the hepatic drug metabolizing enzymes are still unknown. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service