Skip to Content
Merck
All Photos(1)

Key Documents

32273

Supelco

4-tert-Octylphenol-3,5-d2 solution

100 μg/mL in acetone, analytical standard

Synonym(s):

4-(1,1,3,3-Tetramethylbutyl)phenol-3,5-d2, 4-tert-OP-D2

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$891.00

$891.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$891.00

About This Item

Empirical Formula (Hill Notation):
C14D2H20O
CAS Number:
Molecular Weight:
208.34
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

$891.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+2

storage temp.

−20°C

SMILES string

[2H]c1cc(O)cc([2H])c1C(C)(C)CC(C)(C)C

InChI

1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3/i6D,7D

InChI key

ISAVYTVYFVQUDY-QFIQSOQBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service