Skip to Content
Merck
All Photos(2)

Documents

32138

Supelco

Doripenem monohydrate

VETRANAL®, analytical standard

Synonym(s):

(4R,5S,6S)-6-[(1R)-1-Hydroxyethyl]-4-methyl-7-oxo-3-[(3S,5S)-5-[(sulfamoylamino)methyl]pyrrolidin-3-yl]sulfanyl-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N4O6S2 · H2O
CAS Number:
Molecular Weight:
438.52
Beilstein:
15340136
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[H]O[H].NS(NC[C@H](C1)NC[C@H]1SC2=C(C(O)=O)N3C([C@]([H])([C@H](O)C)[C@@]3([H])[C@H]2C)=O)(=O)=O

InChI

1S/C15H24N4O6S2.H2O/c1-6-11-10(7(2)20)14(21)19(11)12(15(22)23)13(6)26-9-3-8(17-5-9)4-18-27(16,24)25;/h6-11,17-18,20H,3-5H2,1-2H3,(H,22,23)(H2,16,24,25);1H2/t6-,7-,8+,9+,10-,11-;/m1./s1

InChI key

NTUBEBXBDGKBTJ-WGLOMNHJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Doripenem belongs to the carbapenems family of β-lactam antibiotic compounds.

Application

Doripenem monohydrate can be used as the precursor for the preparation of doripenem which may be used as an analytical reference standard for the determination of doripenem in:
  • Bovine muscle by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
  • Bulk drug and formulations by high performance thin layer chromatography (HPTLC).
  • Medicinal products by capillary electrophoresis coupled to ultraviolet (UV) detector as well as micellar electrokinetic chromatography (MEKC) with UV detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A validated HPTLC method for the determination of doripenem in pharmaceutical dosage forms.
Kurien J, et al.
International Journal of Pharmaceutical Sciences Review and Research, 27, 317-321 (2014)
Determination of doripenem and related substances in medicinal product using capillary electrophoresis.
Michalska K, et al.
Journal of Separation Science, 34(4), 475-482 (2011)
Determination of biapenem in a medicinal product by micellar electrokinetic chromatography with sweeping in an enhanced electric field.
Michalska K, et al.
Journal of Chromatography A, 1282(4), 153-160 (2013)
Cynthia L Bristow et al.
Frontiers in pharmacology, 12, 695971-695971 (2021-06-29)
The abundant blood protein α1-proteinase inhibitor (α1PI, Αlpha-1, α1-antitrypsin, SerpinA1) is known to bind to the active site of granule-associated human leukocyte elastase (HLE-G). Less well known is that binding of α1PI to cell surface HLE (HLE-CS) induces lymphocyte locomotion
M Di Rocco et al.
Journal of chromatography. A, 1500, 121-135 (2017-04-30)
A method was developed for the confirmatory and quantitative analysis of 30 β-lactam antibiotic residues in bovine muscle. The method includes 12 penicillins (amoxicillin, ampicillin, cloxacillin, dicloxacillin, mecillinam, methicillin, nafcillin, oxacillin, penicillin G, penicillin V, piperacillin, ticarcillin), 12 cephalosporins (cefacetrile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service