Skip to Content
Merck
All Photos(1)

Documents

31577

Supelco

Pyrimethanil

PESTANAL®, analytical standard

Synonym(s):

2-Anilino-4,6-dimethylpyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N3
CAS Number:
Molecular Weight:
199.25
Beilstein:
151589
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(C)nc(Nc2ccccc2)n1

InChI

1S/C12H13N3/c1-9-8-10(2)14-12(13-9)15-11-6-4-3-5-7-11/h3-8H,1-2H3,(H,13,14,15)

InChI key

ZLIBICFPKPWGIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrimethanil is an anilino-pyrimidine, broad spectrum fungicide primarily used for the control of gray mould and leaf scab on fruits, vegetables and ornamentals. Its mode of action involves inhibition of the secretion of hydrolytic enzymes by the fungi during the infection process, thus stopping the further development of the disease.

Application

Pyrimethanil may be used as a reference standard for the determination of the analyte in fruits and ketchup samples by gas chromatography-mass spectrometry in combination with headspace solid-phase microextraction method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangyang Yu et al.
Journal of environmental sciences (China), 22(4), 615-620 (2010-07-14)
Biochar derived from partial combustion of vegetation is ubiquitous and potentially effective in sequestration of environmental contaminants. Biochars were prepared by burning of red gum (Eucalyptus spp.) woodchips at 450 and 850 degrees C (labeled as BC450 and BC850). These
Sarah C Wood et al.
Insects, 11(4) (2020-04-23)
Neonicotinoid and fungicide exposure has been linked to immunosuppression and increased susceptibility to disease in honeybees (Apis mellifera). European foulbrood, caused by the bacterium Melissococcus plutonius, is a disease of honeybee larvae which causes economic hardship for commercial beekeepers, in
Anne Seeland et al.
Environmental pollution (Barking, Essex : 1987), 174, 1-9 (2012-12-19)
It can be suggested that the combined stress of pesticide pollution and suboptimal temperature influences the sensitivity of life stages of aquatic invertebrates differently. The embryo, juvenile, half- and full-life-cycle toxicity tests performed with the snail Physella acuta at different
Anne Seeland et al.
Environmental pollution (Barking, Essex : 1987), 168, 161-169 (2012-05-25)
The aquatic ecotoxic profile of the fungicide pyrimethanil and its acute and chronic thermal dependence in two aquatic invertebrates Chironomus riparius and Daphnia magna were investigated. The ecotoxicity of pyrimethanil at optimal thermal conditions did not depend on the trophic
Luisa Mandrile et al.
Biosensors, 9(4) (2019-12-22)
Flexible and transparent substrates are emerging as low cost and easy-to-operate support for surface-enhanced Raman spectroscopy (SERS). In particular, in situ SERS detection approach for surface characterization in transmission modality can be efficiently employed for non-invasive analysis of non-planar surfaces.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service