Skip to Content
Merck
All Photos(1)

Key Documents

31563

Supelco

2-Methyl-p-phenylenediamine sulfate salt

analytical standard

Synonym(s):

2,5-Toluenediamine sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)2 · H2SO4
CAS Number:
Molecular Weight:
220.25
Beilstein:
3755478
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OS(O)(=O)=O.Cc1cc(N)ccc1N

InChI

1S/C7H10N2.H2O4S/c1-5-4-6(8)2-3-7(5)9;1-5(2,3)4/h2-4H,8-9H2,1H3;(H2,1,2,3,4)

InChI key

KZTWOUOZKZQDMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Skazik et al.
Contact dermatitis, 59(4), 203-211 (2008-10-11)
Patch tests to p-phenylenediamine (PPD) and related substances often show concurrent reactions that can be attributed to separate sensitization or cross-reactivity. In order to understand the health risks associated with cross-reactivity, we studied cross-reactivity of eight chemicals in vitro by
Suhan Gunasti et al.
Indian journal of dermatology, venereology and leprology, 76(2), 165-167 (2010-03-17)
Hair coloring with henna has been popular in Turkey for years. In recent years since the tattoos are applied by the street vendors in most of the beach places in Turkey, skin coloring with henna has also increased. Henna can
Allergic contact dermatitis from toluene-2,5-diamine in a cream dye for eyelashes and eyebrows--quantitative exposure assessment.
Heidi Søsted et al.
Contact dermatitis, 57(3), 195-196 (2007-08-08)
Primary sensitization to toluene-2,5-diamine giving rise to early positive patch reaction to p-phenylenediamine and late to toluene-2,5-diamine.
A Bregnhøj et al.
Contact dermatitis, 59(3), 189-190 (2008-09-02)
Andrew Scheman et al.
Dermatitis : contact, atopic, occupational, drug, 22(4), 189-192 (2011-07-26)
Finding alternative hair dyes for individuals allergic to para-phenylenediamine (PPD) has been difficult. Newer permanent and demipermanent hair dyes that have replaced PPD with para-toluenediamine sulfate (PTDS) are now available. We examined whether individuals allergic to PPD will tolerate PPD-free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service