Skip to Content
Merck
All Photos(2)

Key Documents

31461

Supelco

Carbophenothion

PESTANAL®, analytical standard

Synonym(s):

S-(4-Chlorophenylthiomethyl) O,O-diethyl phosphorodithioate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$75.50

$75.50


Estimated to ship onMay 09, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$75.50

About This Item

Empirical Formula (Hill Notation):
C11H16ClO2PS3
CAS Number:
Molecular Weight:
342.87
Beilstein:
1885240
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$75.50


Estimated to ship onMay 09, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCSc1ccc(Cl)cc1

InChI

1S/C11H16ClO2PS3/c1-3-13-15(16,14-4-2)18-9-17-11-7-5-10(12)6-8-11/h5-8H,3-4,9H2,1-2H3

InChI key

VEDTXTNSFWUXGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Carbophenothion is an organophosphorus pesticide which is highly toxic through ingestion as well as skin absorption. It is mostly used as non-systemic insecticide and acaricide.[1][2]

Application

Carbophenothion may have been used as internal standard for identification and characterisation of organochlorine and organophosphorus pesticides in Brazil′s medicinal plants like Passiflora L. using high-resolution gas chromatography with electron-capture and flame photometric detection (HRGC–ECD/FPD).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast supercritical fluid extraction and high-resolution gas chromatography with electron-capture and flame photometric detection for multiresidue screening of organochlorine and organophosphorus pesticides in Brazil's medicinal plants.
Zuin VG, Yariwake JH, Bicchi C.
Journal of Chromatography A, 985(1-2), 159-166 (2003)
T.S.S. Dikshith
Handbook of Chemicals and Safety, 167-167 (2016)
Dislodgeable residues of carbophenothion in Florida citrus: implications for safe worker reentry.
G C de Batista et al.
Bulletin of environmental contamination and toxicology, 35(2), 213-221 (1985-08-01)
S Sánchez-Fortún et al.
Environmental toxicology and chemistry, 20(9), 2008-2013 (2001-08-28)
The acute toxicity of carbophenothion to three age classes of Artemia salina was evaluated. An increase in toxicity of carbophenothion was found following longer development of A. salina. The effect of pretreatment with the nonselective muscarinic antagonist atropine, the two
Evaluation of organophosphorus pesticide residues in citrus fruits from the Valencian community (Spain).
Torres CM
Journal of AOAC (Association of Official Analytical Chemists) International, 80(5), 1122-1128 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service