Skip to Content
Merck
All Photos(1)

Key Documents

31119-M

Sigma-Aldrich

Ammonium sulfate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12352314
eCl@ss:
38030408
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥99%

form

crystalline

impurities

≤0.005% in water insoluble matters
≤5 ppm heavy metals (by ICP-OES)

ign. residue

≤0.005% (as SO4)

loss

≤0.1% loss on drying, 110 °C

pH

5-6 (25 °C, 5%)

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤3 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg

cation traces

As: ≤0.00002%
Ca: ≤0.001%
Cd: ≤0.0001%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0002%
Fe: ≤2 mg/kg
K: ≤0.005%
Mg: ≤0.0002%
Mn: ≤0.0005%
Na: ≤0.002%
Ni: ≤0.0005%
Pb: ≤0.0002%
Zn: ≤0.0001%

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium sulfate is an inorganic salt that can be produced from the direct neutralization of sulfuric acid with ammonia[1].

Application

Ammonium sulfate can be used as a nucleating agent in the preparation of fatty acid microcapsules[2].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium compounds
Kirk-Othmer Encyclopedia of Chemical Technology (2000)
Microencapsulation of a fatty acid with Poly (melamine-urea-formaldehyde
Konuklu Y, et al.
Energy Conversion and Management , 80, 382-390 (2014)
Separation of proteins by ammonium sulfate gradient solubilization.
T P King
Biochemistry, 11(3), 367-371 (1972-02-01)
Deliquescence and crystallization of ammonium sulfate particles internally mixed with water-soluble organic compounds
Parsons MT, et al.
The Journal of Physical Chemistry A, 108(52), 11600-11608 (2004)
Neutron-Diffraction Study of the Structures of Ferroelectric and Paraelectric Ammonium Sulfate.
Schlemper EO, et al.
J. Chem. Phys. , 44(12), 4498-4509 (1966)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service