Skip to Content
Merck
All Photos(1)

Documents

310441

Sigma-Aldrich

Lead(II) chromate

ACS reagent, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCrO4
CAS Number:
Molecular Weight:
323.19
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

Assay

≥98.0%

impurities

≤0.01% C compounds
≤0.15% solubles

SMILES string

[PbH2++].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.4O.Pb/q;;;2*-1;+2

InChI key

MOUPNEIJQCETIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1A - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Camyre et al.
Toxicological sciences : an official journal of the Society of Toxicology, 97(2), 348-354 (2007-03-16)
Particulate hexavalent chromium ((Cr(VI)) compounds are human lung carcinogens. These compounds induce DNA damage, chromosome aberrations, and concentration-dependent cell death in human and Chinese hamster ovary (CHO) cells. The relationship between Cr(VI)-induced DNA damage and chromosome aberrations is poorly understood.
John Pierce Wise et al.
Mutation research, 517(1-2), 221-229 (2002-05-30)
Hexavalent chromium (Cr(VI)) is a human lung carcinogen. Cr(VI) is a particularly important and dangerous carcinogen, because there is widespread exposure to it both occupationally and to the general public. However, despite the potential for widespread exposure and the fact
X I Gu et al.
Osteoarthritis and cartilage, 20(9), 1011-1019 (2012-05-23)
Recent developments on high resolution micro computed tomography (μCT) allow imaging of soft tissues in small animal joints. Nevertheless, μCT images cannot distinguish soft tissues from synovial fluid due to their similar mass density, limiting the 3D assessment of soft
M Oujja et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 102, 7-14 (2012-12-04)
In this study, the reaction of widely used artist's pigments in raw form to pulsed laser radiation of different wavelengths and pulse duration was investigated. Vermilion, lead chromate and malachite (in the form of pellets) were irradiated using laser pulses
Eliza Grlickova-Duzevik et al.
Toxicological sciences : an official journal of the Society of Toxicology, 92(2), 409-415 (2006-05-23)
Water-insoluble hexavalent chromium compounds are well-established human lung carcinogens. Lead chromate, a model insoluble Cr(VI) compound, induces DNA damage, chromosome aberrations, and dose-dependent cell death in human and Chinese hamster ovary (CHO) cells. The relationship between lead chromate-induced DNA damage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service