Skip to Content
Merck
All Photos(1)

Key Documents

309958

Sigma-Aldrich

Diethyl ether

≥99.9%, spectrophotometric grade, inhibitor-free

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
grade:
spectrophotometric grade
Assay:
≥99.9%
bp:
34.6 °C (lit.)
vapor pressure:
28.69 psi ( 55 °C)
8.59 psi ( 20 °C)
Pricing and availability is not currently available.

Product Name

Diethyl ether, spectrophotometric grade, ≥99.9%, inhibitor-free

grade

spectrophotometric grade

vapor density

2.6 (vs air)

vapor pressure

28.69 psi ( 55 °C)
8.59 psi ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

320 °F

expl. lim.

36.5 %

impurities

≤0.03% water
≤1 ppm peroxides (as H2O2)

evapn. residue

<0.0003%

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 218 nm Amax: 1.0
λ: 250 nm Amax: 0.20
λ: 275 nm Amax: 0.04
λ: 300-400 nm Amax: 0.01

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel L Villeneuve et al.
Ecotoxicology and environmental safety, 68(1), 20-32 (2007-04-24)
An in vitro steroidogenesis assay using H295R human adenocarcinoma cells has been suggested as a possible alternative to gonad explant assays for use as a Tier I screening assay to detect endocrine active chemicals capable of modulating steroid hormone synthesis.
Daniel L Villeneuve et al.
Toxicological sciences : an official journal of the Society of Toxicology, 104(1), 113-123 (2008-04-10)
A number of environmental contaminants and plant flavonoid compounds have been shown to inhibit the activity of 3beta-hydroxysteroid dehydrogenase/Delta(5)-Delta(4) isomerase (3beta-HSD). Because 3beta-HSD plays a critical role in steroid hormone synthesis, inhibition of 3beta-HSD represents a potentially important mode of
Diethyl ether (DEE) as a renewable diesel fuel.
Bailey B, et al.
No. 972978. SAE technical paper (1997)
Oxidation of secondary alcohols in diethyl ether with aqueous chromic acid. Convenient procedure for the preparation of ketones in high epimeric purity.
Brown HC, et al.
The Journal of Organic Chemistry, 36(3), 387-390 (1971)
Eagleson M.
Concise Encyclopedia Chemistry, 471-471 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service