Skip to Content
Merck
All Photos(1)

Key Documents

30909

Supelco

Denaturation solution for in situ hybridization

BioUltra, for molecular biology

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116105

grade

Molecular Biology
for molecular biology

Quality Level

product line

BioUltra

composition

NaCl, 1.5 M
NaOH, 0.5 M

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.352

pH

~13

cation traces

Al: ≤2 mg/kg
Ba: ≤2 mg/kg
Bi: ≤2 mg/kg
Ca: ≤10 mg/kg
Cd: ≤2 mg/kg
Co: ≤2 mg/kg
Cr: ≤2 mg/kg
Cu: ≤2 mg/kg
Fe: ≤2 mg/kg
K: ≤100 mg/kg
Li: ≤2 mg/kg
Mg: ≤2 mg/kg
Mn: ≤2 mg/kg
Mo: ≤2 mg/kg
Ni: ≤2 mg/kg
Pb: ≤2 mg/kg
Sr: ≤2 mg/kg
Zn: ≤2 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.02
λ: 280 nm Amax: 0.01

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Maniatis et al.
Molecular Cloning: A Laboratory Manual (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service