Skip to Content
Merck
All Photos(4)

Documents

307521

Sigma-Aldrich

Resorcinol

ReagentPlus®, 99%

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 21.1 °C)

product line

ReagentPlus®

Assay

99%

form

chips
powder or granules

autoignition temp.

1126 °F

color

white

bp

178 °C/16 mmHg (lit.)
178 °C/16 mmHg

mp

109-112 °C (lit.)

solubility

H2O: 5% (50 mg/ml)

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Resorcinol, a dihydric phenol, is a key component of hair dyeing products. Studies in B16-F10 melanoma cells have revealed that it minimizes melanin production, making it a potential hypopigmenting agent.It serves as a cross-linking agent to produce resorcinol-formaldehyde resin, adhesives, rubbers, and composite materials.

Application

Resorcinol may be used as a starting material in the preparation of organic compounds such as resacetophenone, 4-methyl-7-hydroxycoumarin, β-resorcylic acid and 2-methyl-1,3-cyclohexanedione.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Resacetophenone
Cooper SR
Organic Syntheses, 21, 103-103 (1941)
2-Methyl-1,3-cyclohexanedione
Mekler AB, et al.
Organic Syntheses, 41, 56-56 (1961)
Resorcinol-formaldehyde resins as metal-free semiconductor photocatalysts for solar-to-hydrogen peroxide energy conversion
Y Shiraishi, et al.
Nature Materials, 18, 985-993 (2019)
2,6-Dihydroxyacetophenone
Russell A and Frye JR
Organic Syntheses, 21, 22-22 (1941)
?-Resorcylic acid
Nierenstein M and Clibbens DA
Organic Syntheses, 10, 94-94 (1930)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service