Skip to Content
Merck
All Photos(1)

Key Documents

30648

Supelco

Diallyl disulfide

analytical standard

Synonym(s):

Allyl disulfide

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$189.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$189.00

About This Item

Linear Formula:
CH2=CHCH2SSCH2CH=CH2
CAS Number:
Molecular Weight:
146.27
Beilstein:
1699241
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

refractive index

n20/D 1.539-1.543

density

1.0 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C=CCSSCC=C

InChI

1S/C6H10S2/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-6H2

InChI key

PFRGXCVKLLPLIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diallyl disulfide is an oil-soluble organosulfer compound found in garlic,[1] which is known to exert chemopreventive activity against colon, lung, and skin cancer.[2]

Application

Diallyl disulfide may be used as a reference standard for the determination of diallyl disulfide in human breath samples after ingestion of raw garlic using gas chromatography coupled with mass spectrometry (GC-MS).[3]

Caution

Store under argon.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Odor components of human breath after the ingestion of grated raw garlic.
Minami T, et al.
Journal of Food Science, 54(3), 763-763 (2002)
Induction of apoptosis by diallyl disulfide through activation of caspase-3 in human leukemia HL-60 cells1.
Kwon B-K, et al.
Biochemical Pharmacology, 63(1), 41-47 (2002)
Growth inhibitory effects of diallyl disulfide on human breast cancer cell lines.
Nakagawa H, et al.
Carcinogenesis, 22(6), 891-897 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service