Skip to Content
Merck
All Photos(1)

Key Documents

30608

Supelco

1-Decanol

Selectophore, for ion-selective electrodes, ≥98.0%

Synonym(s):

n-Decyl alcohol, Alcohol C10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
Beilstein:
1735221
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.05

grade

for ion-selective electrodes

Quality Level

vapor density

4.5 (vs air)

vapor pressure

1 mmHg ( 70 °C)
8.25 mmHg ( 100 °C)

product line

Selectophore

Assay

≥98.0% (GC)
≥98.0%

autoignition temp.

550 °F

refractive index

n20/D 1.437 (lit.)

bp

231 °C (lit.)

mp

5-7 °C (lit.)

solubility

H2O: slightly soluble 0.0211 g/L at 20 °C

density

0.829 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCO

InChI

1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3

InChI key

MWKFXSUHUHTGQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - Pensky-Martens closed cup

Flash Point(C)

95 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Sanhita S Dixit et al.
Langmuir : the ACS journal of surfaces and colloids, 26(9), 6193-6200 (2010-04-07)
We demonstrate the optical manipulation of nanoliter aqueous droplets containing surfactant or lipid molecules and immersed in an organic liquid using near-infrared light. The resulting emulsion droplets are manipulated using both the thermocapillary effect and convective fluid motion. Droplet-pair interactions
Anne M Shearrow et al.
Journal of chromatography. A, 1216(36), 6349-6355 (2009-08-01)
Two ionic liquids (IL), namely, 1-methyl-3-octylimidazolium chloride (MOIC) and trihexyltetradecylphosphonium tetrafluoroborate (TTPT) were used to prepare polar and nonpolar sol-gel coatings for capillary microextraction (CME). Bis[(3-methyldimethoxysilyl)propyl] polypropylene oxide (BMPO), containing sol-gel active terminal methoxysilyl groups and polar propylene oxide repeating
Darren J Caven-Quantrill et al.
Journal of chromatography. A, 1218(7), 875-881 (2011-01-11)
A stir bar sorptive extraction (SBSE) method coupled with gas chromatography-mass spectrometry was optimised for the analysis of volatile components of a model wine, based on a previously optimised method used for analysis of the same components in model grape
Armen H Poghosyan et al.
The journal of physical chemistry. B, 113(5), 1303-1310 (2009-01-30)
We have performed a 50 ns of molecular dynamics study of poly(diallyldimethylammonium chloride) (PDADMAC)/sodium dodecyl sulfate (SDS)/decanol/water systems. The influence of the cationic polyelectrolyte on the anionic SDS-based lamellar liquid crystalline system was investigated. The main structural parameters have been
H Ahumada et al.
The journal of physical chemistry. A, 109(30), 6644-6651 (2006-07-13)
Most drugs have to cross cell membranes to reach their final target. A better understanding of the distribution, interactions, and dynamics of biologically active molecules in model bilayers is of fundamental importance in understanding drug functioning and design. 2H NMR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service