Skip to Content
Merck
All Photos(1)

Documents

30164

Supelco

3-Hydroxyvalproic acid, mixture of diastereomers

analytical standard, ≥98.0% (sum od distereomers, GC)

Synonym(s):

3-Hydroxy-2-propylpentanoic acid, 3-Hydroxy-2-propylvaleric acid, 3-OH-VPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O3
CAS Number:
Molecular Weight:
160.21
Beilstein:
2240423
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (sum od distereomers, GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

CCC(O)C(CCC)C(O)=O

InChI

1S/C8H16O3/c1-3-5-6(8(10)11)7(9)4-2/h6-7,9H,3-5H2,1-2H3,(H,10,11)

InChI key

LLPFTSMZBSRZDV-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service