Skip to Content
Merck
All Photos(1)

Documents

30151

Supelco

Shatavarin IV

analytical standard

Synonym(s):

3β,5β,25S)-Spirostan-3-yl O-6-deoxy-α-L-mannopyranosyl-(1→4)-O-[β-D-glucopyranosyl-(1→2)]-β-D-glucopyranoside, Asparanin B, Curillin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H74O17
CAS Number:
Molecular Weight:
887.06
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥85.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@H]1CC[C@@]2(OC1)O[C@H]3C[C@H]4[C@@H]5CC[C@@H]6C[C@H](CC[C@]6(C)[C@H]5CC[C@]4(C)[C@H]3[C@@H]2C)O[C@@H]7O[C@H](CO)[C@@H](O[C@@H]8O[C@@H](C)[C@H](O)[C@@H](O)[C@H]8O)[C@H](O)[C@H]7O[C@@H]9O[C@H](CO)[C@@H](O)[C@H](O)[C@H]9O

InChI

1S/C45H74O17/c1-19-8-13-45(55-18-19)20(2)30-27(62-45)15-26-24-7-6-22-14-23(9-11-43(22,4)25(24)10-12-44(26,30)5)57-42-39(61-41-36(53)34(51)32(49)28(16-46)58-41)37(54)38(29(17-47)59-42)60-40-35(52)33(50)31(48)21(3)56-40/h19-42,46-54H,6-18H2,1-5H3/t19-,20-,21-,22+,23-,24+,25-,26-,27-,28+,29+,30-,31-,32+,33+,34-,35+,36+,37-,38+,39+,40-,41-,42+,43-,44-,45+/m0/s1

InChI key

BCUDKRWNGQAFLF-PJFZGHSASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Shatavarin IV, glycoside of sarsasapogenin, is the major saponin found in the roots of Asparagus racemosus. It is the an important constituent of nutraceuticals and functional foods with anticarcinogenic and antioxidant properties.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Shatavarin IV may be used as a standard for the determination of shatavarin IV extracted from the roots of Asparagus racemosus using high performance thin layer chromatography method (HPTLC).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunomodulatory activity of Asparagus racemosus on systemic Th1/Th2 immunity: implications for immunoadjuvant potential.
Gautam M, et al.
Journal of Ethnopharmacology, 121(2), 241-247 (2009)
Determination of Shatavarin-IV in Asparagus racemosus by high performance thin layer chromatography.
Jain P K, et al.
Journal of Research and Education in Indian Medicine, 15-18 (2009)
Shatavarins (containing Shatavarin IV) with anticancer activity from the roots of Asparagus racemosus.
Mitra KS, et al.
Indian Journal of Pharmacology, 44(6), 732-732 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service