Skip to Content
Merck
All Photos(1)

Key Documents

26402

Sigma-Aldrich

Aluminium distearate

purum, 4.5-6% Al basis

Synonym(s):

Stearic acid aluminum salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$62.30

$62.30


Estimated to ship onMay 07, 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$62.30

About This Item

Empirical Formula (Hill Notation):
C36H71AlO5
CAS Number:
Molecular Weight:
610.93
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.30


Estimated to ship onMay 07, 2025


Request a Bulk Order

Quality Level

grade

purum

form

powder

concentration

4.5-6% Al

impurities

≤0.002% heavy metals (as Pb)
≤7% free acid (as stearic acid)

ign. residue

10-12%

loss

≤1.5% loss on drying, 105 °C, 3 h

solubility

water: insoluble

SMILES string

[Al](OC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC.O

InChI

1S/2C18H36O2.Al.H2O/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;;/h2*2-17H2,1H3,(H,19,20);;1H2/q;;+2;/p-2

InChI key

ZFBYSSBIQZUBBE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Aluminumdisterate is an organic compound generally used as a thickening agent, lubricating agent and water repellent in chemical industry.

Application

Aluminium distearate has been used in the preparation of Aluminum di-soaps[1].
Aluminiumdistearate can be used in the preparation of carbon/alumina nanocomposites from aluminium carboxylate precursors that are used to remove sulphide ions from water.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ushadevi Balasubramani et al.
Journal of hazardous materials, 340, 241-252 (2017-07-18)
An investigation of adsorption of sulphide ion (S
Preparation of Aluminum Di-Soaps1.
Smith GH, et al.
Journal of the American Chemical Society, 70(3), 1053-1054 (1948)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service