Skip to Content
Merck
All Photos(1)

Key Documents

25422

Sigma-Aldrich

Sodium oxalate

puriss., ≥99%

Synonym(s):

Ethanedioic acid sodium salt, Oxalic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

puriss.

Assay

≥99%

impurities

≤0.002% heavy metals (as Pb)

pH

6.5-8.5 (20 °C, 2.5%)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Fe: ≤20 mg/kg

SMILES string

[Na+].[Na+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Joo Lee et al.
Bioresource technology, 130, 211-217 (2013-01-12)
The present study investigated the feasibility of the recovery and reuse oxalic acid in a multistage process for the pretreatment of a lignocellulosic biomass. Electrodialysis (ED), an electrochemical process using ion exchange membranes, was used to recover and reuse oxalic
Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this
F Debela et al.
Journal of environmental management, 116, 156-162 (2013-01-15)
Pyromorphite (PY) and some zinc phosphates (Zn-P) are very sparingly soluble minerals and hence can immobilize Pb and Zn in contaminated soils. However, mechanisms leading to the poor efficiency of PY and Zn-P formation in contaminated soils amended with P
Lars Poulsen et al.
PloS one, 7(12), e50596-e50596 (2012-12-20)
Acid formation in Aspergillus niger is known to be subjected to tight regulation, and the acid production profiles are fine-tuned to respond to the ambient pH. Based on transcriptome data, putative trans-acting pH responding transcription factors were listed and through
Peiyan Li et al.
Food chemistry, 142, 72-78 (2013-09-05)
Effects of oxalic acid on chilling injury, proline metabolism and energy status in mango fruit were investigated after mango fruit (Mangifera indica L. cv. Zill) were dipped in 5mM oxalic acid solution for 10min at 25°C and then stored at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service