Skip to Content
Merck
All Photos(3)

Key Documents

247006

Sigma-Aldrich

5-Sulfosalicylic acid dihydrate

ACS reagent, ≥99%

Synonym(s):

2-Hydroxy-5-sulfobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SC6H3-2-(OH)CO2H·2H2O
CAS Number:
Molecular Weight:
254.21
Beilstein:
650741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥99%
99.0-101.0% (ACS specification)

form

solid

impurities

≤0.02% insolubles
≤0.04% salicylic acid

ign. residue

≤0.1%

mp

105-110 °C (lit.)

solubility

water: soluble 127.1 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.001%
heavy metals: ≤0.002%

functional group

carboxylic acid
sulfonic acid

SMILES string

[H]O[H].[H]O[H].OC(=O)c1cc(ccc1O)S(O)(=O)=O

InChI

1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2

InChI key

BHDKTFQBRFWJKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Sulfosalicylic acid dihydrate is a poly functional metal chelating ligand that may be used to form metal coordination complexes.
Spray reagent for the detection of amino acids on TLC plates. Doping agent for polyaniline.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and physical properties of calcium and strontium complexes with the ligand sulfosalicylic acid
Wu G, et al
Zeitschrift fur Anorganische und Allgemeine Chemie, 636(3?4), 652-656 (2010)
Mohammad Habibi et al.
Animal nutrition (Zhongguo xu mu shou yi xue hui), 7(3), 868-882 (2021-10-12)
The objective of this study was to assess the growth efficiency of pigs fed with protein-restricted diets supplemented with branched-chain amino acids (BCAA) and limiting amino acids (LAA) above the recommended levels. Following 2 weeks of adaptation, 48 young barrows were
Synthetic Metals, 58, 309-309 (1993)
Charge transport and optical and magnetic properties of polyaniline synthesized with use of organic acids.
Raghunathan et al.
Physical review. B, Condensed matter, 47(20), 13189-13196 (1993-05-15)
Butyltin (IV) 5-sulfosalicylates: structural characterization and their cytostatic activity
Pruchnik H, et al
Polyhedron, 49(1), 223-233 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service