Skip to Content
Merck
All Photos(3)

Key Documents

234087

Sigma-Aldrich

Potassium indigotrisulfonate

ozone-scavenging reagent

Synonym(s):

5,5′,7-Indigotrisulfonic acid tripotassium salt, Indigo-5,5′,7-trisulfonic acid tripotassium salt, Potassium indigotrisulfonate, Tripotassium indigo-5,5′,7-trisulfonate, tri-Potassium indigo-5,5′,7-trisulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$189.00
5 G
$683.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$189.00
5 G
$683.00

About This Item

Empirical Formula (Hill Notation):
C16H7K3N2O11S3
CAS Number:
Molecular Weight:
616.72
Beilstein:
4932187
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$189.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

form

crystals or chunks

concentration

>60% (HPLC)

technique(s)

titration: suitable

solubility

water: 0.1%

λmax

600 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[K+].[K+].[K+].[O-]S(=O)(=O)c1ccc2NC(\C(=O)c2c1)=C3\Nc4c(cc(cc4S([O-])(=O)=O)S([O-])(=O)=O)C3=O

InChI

1S/C16H10N2O11S3.3K/c19-15-8-3-6(30(21,22)23)1-2-10(8)17-13(15)14-16(20)9-4-7(31(24,25)26)5-11(12(9)18-14)32(27,28)29;;;/h1-5,17-18H,(H,21,22,23)(H,24,25,26)(H,27,28,29);;;/q;3*+1/p-3/b14-13+;;;

InChI key

XOSMXDUITYWYGR-VPOCKCTCSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Ozone-scavenging reagent that reacts rapidly with ozone.[1]
Potassium indigotrisulfonate has been used:
  • in colorimetric method of measurement of atmospheric ozone[2]
  • for measuring residual aqueous ozone by gas diffusion flow injection analysis.[3]
  • to determine O3 concentration in water.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chlorophylls Reductions in Fresh-Cut Chard (Beta vulgaris var. cicla) with Various Sanitizing Agents
Karaca H
Journal of Agricultural Science, 22(2016), 9-19 (2014)
Samira Nahim-Granados et al.
Water research, 170, 115304-115304 (2019-12-02)
In this research, the capability of ozonation and peroxone treatment for the simultaneous disinfection and decontamination of wash water from the fresh-cut industry has been investigated at pilot plant scale (10 L). The removal efficiency of six organic microcontaminants (OMCs) (four
Ozone-scavenging reagents suitable for use in the quantitative determination of aldehydes as the O-(2,3,4,5,6-pentafluorobenzyl) oximes by GC-ECD
Urbansky, E.T., et al.
Water Research, 34, 2610-2613 (2000)
Jingxin Yang et al.
Water research, 176, 115751-115751 (2020-04-06)
Thiosulfate (S2O32-) is frequently used as an ozone (O3) quenching agent when investigating the ozonation of organic contaminants and the kinetics thereof. In this study, however, O3 is activated by S2O32-, resulting in a superfast degradation of O3-refractory contaminants. Therefore
Colorimetric determination of ambient ozone using indigo blue droplet.
Felix, E.P. and Cardoso, A.A.
Journal of the Brazilian Chemical Society, 17, 296-301 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service