Skip to Content
Merck
All Photos(2)

Key Documents

22308

Supelco

Ponceau Xylidine

analytical standard

Synonym(s):

Acid Red 26, Ponceau 2 R, Xylidine ponceau, Xylidine ponceau 2R

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$255.00

$255.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$255.00

About This Item

Empirical Formula (Hill Notation):
C18H14N2Na2O7S2
CAS Number:
Molecular Weight:
480.42
Colour Index Number:
16150
Beilstein:
5709350
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$255.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].Cc1ccc(\N=N\c2c(O)c(cc3cc(ccc23)S([O-])(=O)=O)S([O-])(=O)=O)c(C)c1

InChI

1S/C18H16N2O7S2.2Na/c1-10-3-6-15(11(2)7-10)19-20-17-14-5-4-13(28(22,23)24)8-12(14)9-16(18(17)21)29(25,26)27;;/h3-9,21H,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2/b20-19+;;

InChI key

YJVBLROMQZEFPA-LLIZZRELSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ponceau xylidine is a synthetic food additive dye widely used in foodstuffs to restore the natural food color and also to enhance food appeal.[1]

Application

Ponceau xylidine may be used as a reference standard for its determination in animal feed and meat using high performance liquid chromatography-diode array detection (HPLC-DAD) and tandem mass spectrometry (HPLC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of seven synthetic dyes in animal feeds and meat by high performance liquid chromatography with diode array and tandem mass detectors.
Zou T, et al.
Food Chemistry, 138(2-3), 1742-1748 (2013)
Shota Kurotsu et al.
The Journal of pharmacology and experimental therapeutics, 350(1), 79-88 (2014-04-29)
Idiopathic pulmonary fibrosis is thought to involve lung injury caused by reactive oxygen species (ROS), which in turn is followed by abnormal fibrosis. A transforming growth factor (TGF)-β1-induced increase in myofibroblast number plays an important role in this abnormal fibrosis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service