Skip to Content
Merck
All Photos(1)

Key Documents

21120

Supelco

Calcium citrate tribasic tetrahydrate

≥98.0% (calc. on dry substance, KT)

Synonym(s):

Citric acid calcium salt, Tricalcium dicitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca3(C6H5O7)2 · 4H2O
CAS Number:
Molecular Weight:
570.49
Beilstein:
3924520
EC Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Assay

≥98.0% (calc. on dry substance, KT)

form

powder

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤150 mg/kg

cation traces

Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤200 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.OC1(CC(=O)O[Ca]OC(=O)C1)C(=O)O[Ca]OC(=O)C2(O)CC(=O)O[Ca]OC(=O)C2

InChI

1S/2C6H8O7.3Ca.4H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;4*1H2/q;;3*+2;;;;/p-6

InChI key

LNIZKKFWMDARJV-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alicia Lagarto et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 65(1-2), 49-53 (2011-06-28)
The potential of oral exposure to calcium and magnesium citrate, a natural product obtained from dolomite, to initiate teratogenesis was analyzed in Wistar rats. Animals received calcium and magnesium citrate oral doses of 250, 500 and 1000 mg/kg during the
José Manuel Quesada Gómez et al.
Clinical drug investigation, 31(5), 285-298 (2011-03-17)
The combination of calcium with vitamin D (vitamin D(3) [colecalciferol]) forms the basis of preventive and therapeutic regimens for osteoporosis. A number of studies have suggested that the combination of calcium and vitamin D is effective when administered at respective
Morton G Burt et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 28(2), 412-418 (2012-09-20)
Calcium supplements have been associated with an increased risk of cardiovascular events. However, the validity of these findings has been questioned. A major concern is that the mechanism underlying an increase in cardiovascular events has not been demonstrated. Calcium initiates
Isabelle Zamfirescu et al.
Thyroid : official journal of the American Thyroid Association, 21(5), 483-486 (2011-05-21)
Calcium carbonate is a commonly used dietary supplement and has been shown to interfere with levothyroxine absorption. However, calcium citrate, which is also used for supplementation purposes, has not been studied previously and calcium acetate, which is used to treat
Sherry Huang et al.
Nanotechnology, 20(37), 375102-375102 (2009-08-27)
Taking calcium supplements can reduce the risk of developing osteoporosis, but they are not readily absorbed in the gastrointestinal tract. Nanotechnology is expected to resolve this problem. In the present study, we examined whether the bioavailability of calcium carbonate and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service