Skip to Content
Merck
All Photos(1)

Key Documents

21098

Sigma-Aldrich

Calcium chloride dihydrate

BioUltra, ≥99.5% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

product line

BioUltra

Assay

≥99.5% (KT)

form

solid

impurities

Insoluble matter, passes filter test
≤0.002% Total nitrogen (N)

pH

5.0-8.0 (25 °C, 1 M in H2O)

mp

176 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤20 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤100 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.011
λ: 280 nm Amax: 0.010

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Constituent of cell culture media. Preparation of microsomes by calcium aggregation; Preparation of mitochondrial ribosomes in which Ca2+ inhibits nuclease activity during mitochondrial lysis; Preparation of calcium alginate gels for microbial cell immobilization by gel entrapment; Role of calcium in the adhesion and fusion of bilayers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of microsomes with calcium.
J B Schenkman et al.
Methods in enzymology, 52, 83-89 (1978-01-01)
M Kierstan et al.
Biotechnology and bioengineering, 19(3), 387-397 (1977-03-01)
Saccharomyces cerevisiae cells, Kluyveromyces marxianus cells, inulase, glucose oxidase, chloroplasts, and mitochondria were immobilized in calcium alginate gels. Ethanol production from glucose solutions by an immobilized preparation of S. cerevisiae was deomonstrated over a total of twenty-three days, and the
D E Leckband et al.
Biochemistry, 32(4), 1127-1140 (1993-02-02)
The interaction forces and fusion mechanisms of mixed zwitterionic-anionic phospholipid bilayers were measured with the surface forces apparatus. The bilayers were 3:1 mixtures of either dimyristoylphosphatidylcholine and dimyristoylphosphatidylglycerol (DMPC/DMPG) or dilauroylphosphatidylcholine and dilauroylphosphatidylglycerol (DLPC/DLPG), and experiments were carried out in
Preparation and analysis of mitochondrial ribosomes.
A M Lambowitz
Methods in enzymology, 59, 421-433 (1979-01-01)
Stephanie Saul et al.
Journal of leukocyte biology, 106(5), 1101-1115 (2019-06-20)
Neutrophils play a central role in immunity and inflammation via their intrinsic ability to migrate into inflamed tissue, to phagocytose pathogens, and to kill bacterial and fungi by releasing large quantities of superoxide anions and lytic enzymes. The molecular pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service