Skip to Content
Merck
All Photos(1)

Documents

210374

Sigma-Aldrich

Acid Yellow 42

Dye content 25 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H24N8Na2O8S2
CAS Number:
Molecular Weight:
758.69
Colour Index Number:
22910
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 25%

mp

263 °C (dec.) (lit.)

λmax

410 nm

SMILES string

[Na+].[Na+].CC1=NN(C(=O)C1\N=N\c2ccc(-c3ccc(cc3S([O-])(=O)=O)\N=N\C4C(C)=NN(C4=O)c5ccccc5)c(c2)S([O-])(=O)=O)c6ccccc6


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service