20906-U
Hamilton® syringe, 900 series
95N, volume 5 μL, needle size 26s ga (bevel tip), needle L 51 mm (2 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Recommended Products
material
stainless steel needle (point style 2)
needle size
26s ga (bevel tip)
description
Microliter™
feature
needle type N (cemented)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 87920
needle L
51 mm (2 in.)
volume
5 μL
Looking for similar products? Visit Product Comparison Guide
General description
Economical version of 800 series syringes.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microliter is a trademark of Hamilton Co.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service