20779
Hamilton® syringe, 700 series, fixed needle
701N, volume 10 μL, needle size 26s ga (bevel tip), needle L 51 mm (2 in.), pkg of 6 ea
About This Item
Recommended Products
material
stainless steel needle (point style 2)
needle size
26s ga (bevel tip)
feature
needle type N (cemented needle)
point style 2
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 80366
needle L
51 mm (2 in.)
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
General description
700 Series Six Packs
The most popular syringes are available in convenient multi-unit packages, at considerable cost-savings. Accurate to ±1%. Temperature limit: 50° C for cemented needles, 115° C for removable needle.
10μL capacity, 26s gauge, 51mm length, point style #2, in packs of 6.
Application
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service