Skip to Content
Merck
All Photos(1)

Key Documents

20760

Sigma-Aldrich

Butyronitrile

purum, ≥99.0% (GC)

Synonym(s):

Propyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$88.40
1 L
$130.00

$88.40


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$88.40
1 L
$130.00

About This Item

Linear Formula:
CH3CH2CH2CN
CAS Number:
Molecular Weight:
69.11
Beilstein:
1361452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$88.40


Estimated to ship onMay 19, 2025


Request a Bulk Order

vapor density

2.4 (vs air)

Quality Level

vapor pressure

23 mmHg ( 25 °C)

grade

purum

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

910 °F

refractive index

n20/D 1.384 (lit.)
n20/D 1.384

bp

115-117 °C (lit.)

mp

−112 °C (lit.)

solubility

DMF: miscible(lit.)
alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
water: very slightly soluble(lit.)

density

0.794 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

CCCC#N

InChI

1S/C4H7N/c1-2-3-4-5/h2-3H2,1H3

InChI key

KVNRLNFWIYMESJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Molar volumes of vapor-deposited butyronitrile glasses have been studied as a function of deposition temperature[1]. Reactions of butyronitrile with high-temperature water at 330°C and pressures ranging from 128 to 2600bar were investigated[2].

Application

Butyronitrile was used in electrolyte composition in dye-sensitized solar cells†.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stéphanie Claudinot et al.
Nature communications, 11(1), 5645-5645 (2020-11-08)
The formation of hair follicles, a landmark of mammals, requires complex mesenchymal-epithelial interactions and it is commonly believed that embryonic epidermal cells are the only cells that can respond to hair follicle morphogenetic signals in vivo. Here, we demonstrate that
Effect of pressure on the rate of butyronitrile hydrolysis in high-temperature water.
Iyer SD and Klein MT.
Journal of Supercritical Fluids, 10(3), 191-200 (1997)
Frédéric Sauvage et al.
Journal of the American Chemical Society, 133(33), 13103-13109 (2011-06-28)
We elaborated a new electrolyte composition, based on butyronitrile solvent, that exhibits low volatility for use in dye-sensitized solar cells. The strong point of this new class of electrolyte is that it combines high efficiency and excellent stability properties, while
Hideyuki Nakayama et al.
The journal of physical chemistry. B, 117(35), 10311-10319 (2013-08-08)
Molar volumes (Vm) of vapor-deposited ethylcyclohexane (ECH) and butyronitrile (BN, sometimes called butanenitrile) glasses were studied as a function of deposition temperature (Td). ECH glasses deposited at Td sufficiently below their glass-transition temperature (Tg) exhibited changes in Vm on heating
E G DeMaster et al.
Biochemical pharmacology, 46(1), 117-123 (1993-07-06)
n-Butyraldoxime (n-BO) is known to cause a disulfiram/ethanol-like reaction in humans, a manifestation of the inhibition of hepatic aldehyde dehydrogenase (AIDH). As with a number of other in vivo inhibitors of AIDH, n-BO does not inhibit purified AIDH in vitro

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service