Key Documents
20704
Hamilton® syringe, luer tip
750LT, volume 0.5 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Pricing and availability is not currently available.
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80801
needle L
, n/a
volume
0.5 mL
compatibility
for use with Hamilton 750LT (Luer tip)
Related Categories
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Legal Information
Hamilton is a registered trademark of Hamilton Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)