Skip to Content
Merck
All Photos(1)

Key Documents

20251

Sigma-Aldrich

N-Butylmethylamine

purum, ≥98.0% (GC)

Synonym(s):

N-Methylbutylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH(CH2)3CH3
CAS Number:
Molecular Weight:
87.16
Beilstein:
1209231
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.3995 (lit.)
n20/D 1.401

bp

90.5-91.5 °C (lit.)

density

0.736 g/mL at 25 °C (lit.)

SMILES string

CCCCNC

InChI

1S/C5H13N/c1-3-4-5-6-2/h6H,3-5H2,1-2H3

InChI key

QCOGKXLOEWLIDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service