Skip to Content
Merck
All Photos(3)

Key Documents

18503

Supelco

Methyl trifluoromethanesulfonate

for GC derivatization, LiChropur, 98.0%

Synonym(s):

Methyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO2OCH3
CAS Number:
Molecular Weight:
164.10
Beilstein:
774772
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥98.0% (GC)
98.0%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.326 (lit.)
n20/D 1.327

bp

94-99 °C (lit.)

density

1.45 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COS(=O)(=O)C(F)(F)F

InChI

1S/C2H3F3O3S/c1-8-9(6,7)2(3,4)5/h1H3

InChI key

OIRDBPQYVWXNSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl trifluoromethanesulfonate (Methyl triflate) is a strong methylating agent. It is also a powerful alkylating agent and a strong irritant.[1]
Shown to be the most effective monomethylating agent in reactions with potassium enolates.
Useful methylation reagent for the conversion of amines to methyl ammonium triflates.

Application

Methyl trifluoromethanesulfonate was used in the synthesis of 2-Benzyloxy-1-methylpyridinium trifluoromethanesulfonate.[2]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. L. F. Armarego, Christina Li Lin Chai
Purification of Laboratory Chemicals, 189-189 (2013)
S E Snyder et al.
Nuclear medicine and biology, 25(8), 751-754 (1998-12-24)
Synthesis of 1-[11C]methylpiperidin-4-yl propionate ([11C]PMP), an in vivo substrate for acetylcholinesterase, is reported. An improved preparation of 4-piperidinyl propionate (PHP), the immediate precursor for radiolabeling, was accomplished in three steps from 4-hydroxypiperidine by (a) protection of the amine as the
Convenient gas phase bromination of [11C]methane and production of [11C]methyl triflate.
B H Mock et al.
Nuclear medicine and biology, 26(4), 467-471 (1999-06-26)
Q H Zheng et al.
Nuclear medicine and biology, 23(8), 981-986 (1996-11-01)
The important radiotracer precursor 2 beta-carbomethoxy-3 beta-(4-idophenyl)-tropane (beta-CIT, RTI-55) was made in 52% overall yield from cocaine. Key steps were improved conjugate Grignard addition to anhydroecgonine methyl ester with > 3.5:1 2 beta: 2 alpha-isomer selectivity, and a mild new
Comparison of [11C]methyl triflate and [11C]methyl iodide in the synthesis of PET radioligands such as [11C]beta-CIT and [11C]beta-CFT.
K Någren et al.
Nuclear medicine and biology, 22(8), 965-979 (1995-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service