Skip to Content
Merck
All Photos(1)

Key Documents

17151

Millipore

Kanamycin Esculin Azide Agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12 g/L
casein enzymic hydrolysate, 20 g/L
esculin, 1 g/L
ferric ammonium citrate, 0.5 g/L
kanamycin sulfate, 0.02 g/L
sodium azide, 0.15 g/L
sodium chloride, 5 g/L
sodium citrate, 1 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.0±0.2 (25 °C)

application(s)

environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Enterococcus spp.
selective and differential for Streptococcus spp.

Application

For isolation of Group D Streptococci in foodstuffs.
Kanamycin Esculin Azide Agar may be used as a media to obtain predominant lactic acid bacteria associated with fermenting cassava.[1]

Preparation Note

Suspend 44.67 g in 1 litre distilled water. Heat to boiling to dissolve the medium completely. Dispense as desired. Sterilize by autoclaving at 121°C for 15 minutes. Warning: Sodium azide has a tendency to form explosive metal azides with plumbing materials. It is advisable to use enough water to flush off the disposables.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterisation and biochemical properties of predominant lactic acid bacteria from fermenting cassava for selection as starter cultures.
Kostinek M, et al.
International Journal of Food Microbiology, 114(3), 342-351 (2007)

Articles

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service