Skip to Content
Merck
All Photos(1)

Key Documents

16172

Sigma-Aldrich

SODOSIL RA 13

highly alkaline cleaner

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
$206.00

$206.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
5 KG
$206.00

About This Item

UBA Number:
08050038
UNSPSC Code:
12352100
NACRES:
NA.22

$206.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Quality Level

form

solid

impurities

alkalic reac. substances, complies

General description

SODOSIL is alkaline phosphate-free detergent[1].

Application

SODOSIL was used in cleaning plastic ware during determination of lead in whole blood specimens[1]. It was used in rinsing epoxy-coated surfaces[2] and multielectrode arrays[3].

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shimshon Jacobi et al.
Journal of neurophysiology, 97(5), 3597-3606 (2007-03-09)
We investigate the propagation of neural activity along one-dimensional rat hippocampal cultures patterned in lines over multielectrode arrays. Activity occurs spontaneously or is evoked by local electrical or chemical stimuli, with different resulting propagation velocities and firing rate amplitudes. A
G Aspenes et al.
Journal of colloid and interface science, 343(2), 529-536 (2009-12-29)
The mechanisms by which hydrates deposit in a petroleum production line are related to pipeline surface properties, fluid composition and properties, and water cut. In this work, adhesion forces between cyclopentane hydrates and solid surfaces were investigated as a function
Determination of lead in whole blood by graphite furnace atomic absorption spectrometry with matrix modification.
Navarro JA, et al.
Journal of Analytical Atomic Spectrometry, 4(5), 401-406 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service