Skip to Content
Merck
All Photos(3)

Key Documents

15411

Supelco

Boc-Cys-OH

for chiral derivatization, LiChropur, ≥98.5%

Synonym(s):

Boc-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO4S
CAS Number:
Molecular Weight:
221.27
Beilstein:
2450705
MDL number:
UNSPSC Code:
41116105
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

product line

ChiraSelect

Assay

≥98.5%

form

solid

optical activity

[α]20/D +8.0±1.5°, c = 1% in ethanol

optical purity

enantiomeric ratio: ≥99.5:0.5

quality

LiChropur

technique(s)

HPLC: suitable

SMILES string

CC(C)(C)OC(=O)N[C@@H](CS)C(O)=O

InChI

1S/C8H15NO4S/c1-8(2,3)13-7(12)9-5(4-14)6(10)11/h5,14H,4H2,1-3H3,(H,9,12)(H,10,11)/t5-/m0/s1

InChI key

ATVFTGTXIUDKIZ-YFKPBYRVSA-N

General description

Boc-L-cysteine (Boc-Cys-OH) is a high performance liquid chromatography (HPLC) grade chromatographic solvent.

Application

Boc-L-cysteine (Boc-Cys-OH) can may be used as a chromatographic solvent, in an experimental study done to study the biosynthesis of S-(3-hexan-1-ol)-glutathione (3MH-S-glut) and S-(3-hexan-l-ol)-L-cysteine (3MH-S-cys), which act as flavour precursors in wines, in Vitis vinifera grapes exposed to various environmental stress.

Other Notes

Chiral derivatizing agent used together with OPA for assaying the enantiomeric purity of amino acids

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gantumur Battogtokh et al.
Nanomedicine : nanotechnology, biology, and medicine, 18, 315-325 (2018-11-06)
Photodynamic therapy is a clinically approved treatment approach for cancer. However, it has limited applications owing to poor water solubility and the short wavelength absorption of the photosensitizer (PS). We selected a near-infrared photosensitizer, SiNC, and encapsulated into a gold
Oded Bodner et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(34), 6489-6502 (2020-07-15)
D-serine is a physiologic coagonist of NMDA receptors (NMDARs) required for synaptic plasticity, but mechanisms that terminate D-serine signaling are unclear. In particular, the identity of unidirectional plasma membrane transporters that mediate D-serine reuptake has remained elusive. We report that
Reduction-responsive polymeric micelles for anticancer drug delivery.
Xulin Jiang et al.
Journal of controlled release : official journal of the Controlled Release Society, 152 Suppl 1, e36-e37 (2011-12-27)
A Hashimoto et al.
Journal of chromatography, 582(1-2), 41-48 (1992-11-06)
The concurrent determination of free amino acid enantiomers and non-chiral amino acids in rat brain and serum was accomplished by high-performance liquid chromatography with fluorimetric detection after derivatization with N-tert.-butyloxycarbonyl-L-cysteine and o-phthaldialdehyde. The method revealed the presence of a large
Ryushi Kawakami et al.
Journal of bioscience and bioengineering, 124(1), 23-27 (2017-03-28)
A novel amino acid racemase with broad substrate specificity (BAR) was recently isolated from the hyperthermophilic archaeon Pyrococcus horikoshii OT-3. Characterization of this enzyme has been difficult, however, because the recombinant enzyme is produced mainly as an inclusion body in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service